missing translation for 'onlineSavingsMsg'
Learn More

6-tert-Butyl-o-cresol 99.0+%, TCI America™

Catalog No. B072825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B072825G 25 g
B0728500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B072825G Supplier TCI America Supplier No. B072825G
Only null left

Chemical Identifiers

CAS 2219-82-1
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00002239
InChI Key BKZXZGWHTRCFPX-UHFFFAOYSA-N
Synonym 2-tert-Butyl-6-methylphenol
PubChem CID 16678
IUPAC Name 2-tert-butyl-6-methylphenol
SMILES CC1=CC=CC(=C1O)C(C)(C)C

Specifications

Quantity 25 g
UN Number 2430
Formula Weight 164.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-tert-Butyl-o-cresol
EINECSNumber (3)-0521
RTECSNumber SK1578200
TSCA Yes