missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 6-TAMRA, SE (6-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer

Catalog No. C6123
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6123 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6123 Supplier Invitrogen™ Supplier No. C6123

6-TAMRA, SE (6-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer

The isomeric 6-TAMRA, SE is an amine-reactive form of tetramethylrhodamine. The single-isomer preparations of TAMRA are most important for high-resolution techniques such as DNA sequencing and capillary electrophoresis. 6-TAMRA is one of the traditional fluorophores used for automated DNA sequencing.

Specifications

Quantity 5 mg
Product Type 6-TAMRA SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.