missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 6-ROX, SE (6-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

Catalog No. C6126
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6126 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6126 Supplier Invitrogen™ Supplier No. C6126

6-ROX, SE (6-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

The isomeric 6-ROX, SE is an amine-reactive form of carboxy-X-rhodamine and is widely used for oligonucleotide labeling and automated DNA sequencingapplications.

Specifications

Quantity 5 mg
Product Type 6-ROX SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye ROX
Label Type Classic Dyes
What is the excitation and emission wavelength for rhodamine?

Rhodamine is a generic term for a wide variety of cationic dyes whose fluorescence emission can range from green, orange to red. The table below lists the excitation and emission maxima (nm), as well as molar extinction coefficients (“EC”; cm-1 M-1), for various rhodamine dyes (data derived with dye dissolved in methanol).

Dye Excitation Emission EC
Rhodamine B 568 583 88,000
Rhodamine 123 507 529 101,000
Rhodamine 110 499 521 92,000
Rhodamine 6G 528 551 105,000
XRITC 572 596 92,000

For Research Use Only. Not for use in diagnostic procedures.