missing translation for 'onlineSavingsMsg'
Learn More

6-Quinoxalinecarboxylic acid, 95%, Thermo Scientific™

Chemical Identifiers

CAS 6925-00-4
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
InChI Key JGQDBVXRYDEWGM-UHFFFAOYSA-N
Synonym 6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
PubChem CID 674813
IUPAC Name quinoxaline-6-carboxylic acid
SMILES C1=CC2=NC=CN=C2C=C1C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC11701EE
View Documents
N/A 50g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

6925-00-4
174.159
6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
quinoxaline-6-carboxylic acid
C9H6N2O2
JGQDBVXRYDEWGM-UHFFFAOYSA-N
674813
C1=CC2=NC=CN=C2C=C1C(=O)O

Specifications

6925-00-4
C9H6N2O2
JGQDBVXRYDEWGM-UHFFFAOYSA-N
quinoxaline-6-carboxylic acid
674813
95%
95%
6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
C1=CC2=NC=CN=C2C=C1C(=O)O
174.159
174.16
6-Quinoxalinecarboxylic acid
Videos
SDS
Documents

Documents