missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Quinoxalinecarboxylic acid, 95%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 50g |
| Formula Weight | 174.16 |
| Percent Purity | 95% |
| Chemical Name or Material | 6-Quinoxalinecarboxylic acid |