missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Quinolinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America Q007725G
Specifications
| 6-Quinolinecarboxylic Acid | |
| 291°C | |
| C10H7NO2 | |
| 25 g | |
| VXGYRCVTBHVXMZ-UHFFFAOYSA-N | |
| quinoline-6-carboxylic acid | |
| 82571 | |
| ≥98.0% (HPLC,T) |
| 10349-57-2 | |
| Yellow | |
| MFCD00047613 | |
| 6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide | |
| C1=CC2=C(C=CC(=C2)C(=O)O)N=C1 | |
| 173.171 | |
| 173.17 | |
| Crystalline Powder |
Chemical Identifiers
| 10349-57-2 | |
| 173.171 | |
| VXGYRCVTBHVXMZ-UHFFFAOYSA-N | |
| 82571 | |
| C1=CC2=C(C=CC(=C2)C(=O)O)N=C1 |
| C10H7NO2 | |
| MFCD00047613 | |
| 6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide | |
| quinoline-6-carboxylic acid |
Safety and Handling
TSCA : Yes