missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarboxylic Acid 98.0+%, TCI America™

Catalog No. Q00775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00775G 5 g
Q007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00775G Supplier TCI America Supplier No. Q00775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10349-57-2
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00047613
InChI Key VXGYRCVTBHVXMZ-UHFFFAOYSA-N
Synonym 6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide
PubChem CID 82571
IUPAC Name quinoline-6-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)N=C1

Specifications

Melting Point 291°C
Color Yellow
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Quinolinecarboxylic Acid