missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Quinolinecarboxaldehyde 98.0+%, TCI America™
$205.05 - $721.40
Chemical Identifiers
| CAS | 4113-04-6 |
|---|---|
| Molecular Formula | C10H7NO |
| Molecular Weight (g/mol) | 157.17 |
| MDL Number | MFCD00805836 |
| InChI Key | VUAOIXANWIFYCU-UHFFFAOYSA-N |
| Synonym | 6-Formylquinoline |
| PubChem CID | 765653 |
| IUPAC Name | quinoline-6-carbaldehyde |
| SMILES | C1=CC2=C(C=CC(=C2)C=O)N=C1 |
Chemical Identifiers
| 4113-04-6 | |
| 157.17 | |
| VUAOIXANWIFYCU-UHFFFAOYSA-N | |
| 765653 | |
| C1=CC2=C(C=CC(=C2)C=O)N=C1 |
| C10H7NO | |
| MFCD00805836 | |
| 6-Formylquinoline | |
| quinoline-6-carbaldehyde |
Specifications
| 4113-04-6 | |
| Yellow | |
| MFCD00805836 | |
| 6-Formylquinoline | |
| C1=CC2=C(C=CC(=C2)C=O)N=C1 | |
| 157.17 | |
| 157.17 | |
| Crystalline Powder |
| 76°C | |
| C10H7NO | |
| 1 g | |
| VUAOIXANWIFYCU-UHFFFAOYSA-N | |
| quinoline-6-carbaldehyde | |
| 765653 | |
| ≥98.0% (GC,T) | |
| 6-Quinolinecarboxaldehyde |
Safety and Handling
TSCA : No