CAS RN 4113-04-6
IUPAC Name:
quinoline-6-carbaldehyde
Synonyms:
quinoline-6-carbaldehyde
Molecular Weight (g/mol):
157.17
Molecular Formula:
C10H7NO
InChi Key:
VUAOIXANWIFYCU-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C2N=CC=CC2=C1
1
–
4
of
4
results
3
6-Quinolinecarbaldehyde, 97%, Thermo Scientific™
CAS: 4113-04-6 Molecular Formula: C10H7NO Molecular Weight (g/mol): 157.17 InChI Key: VUAOIXANWIFYCU-UHFFFAOYSA-N Synonym: quinoline-6-carbaldehyde,6-quinolinecarboxaldehyde,6-quinolinecarbaldehyde,quinoline-6-carboxaldehyde,6-formylquinoline,6-quinolinemethanal,pubchem10181,quinolin-6-carbaldehyde,acmc-2097el PubChem CID: 765653
4
6-Quinolinecarboxaldehyde 98.0+%, TCI America™
CAS: 4113-04-6 Molecular Formula: C10H7NO Molecular Weight (g/mol): 157.17 MDL Number: MFCD00805836 InChI Key: VUAOIXANWIFYCU-UHFFFAOYSA-N Synonym: 6-Formylquinoline PubChem CID: 765653 IUPAC Name: quinoline-6-carbaldehyde SMILES: C1=CC2=C(C=CC(=C2)C=O)N=C1