missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarboxaldehyde 98.0+%, TCI America™

Catalog No. Q00971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00971G 1 g
Q00975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00971G Supplier TCI America Supplier No. Q00971G
Only null left

Chemical Identifiers

CAS 4113-04-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00805836
InChI Key VUAOIXANWIFYCU-UHFFFAOYSA-N
Synonym 6-Formylquinoline
PubChem CID 765653
IUPAC Name quinoline-6-carbaldehyde
SMILES C1=CC2=C(C=CC(=C2)C=O)N=C1

Specifications

Melting Point 76°C
Color Yellow
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Quinolinecarboxaldehyde