missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarbaldehyde, 97%, Thermo Scientific™

$135.04 - $135.04

Chemical Identifiers

CAS 4113-04-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
InChI Key VUAOIXANWIFYCU-UHFFFAOYSA-N
Synonym quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
PubChem CID 765653
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC04504DA
SDS View Documents
Thermo Scientific Chemicals
CC04504DA
1 g Amber Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4113-04-6
157.17
quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
C10H7NO
VUAOIXANWIFYCU-UHFFFAOYSA-N
765653

Specifications

4113-04-6
Amber Glass Bottle
1 g
VUAOIXANWIFYCU-UHFFFAOYSA-N
765653
97%
97%
C10H7NO
quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
157.17
157.17
6-Quinolinecarbaldehyde
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'