missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarbaldehyde, 97%, Thermo Scientific™

Catalog No. CC04504DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC04504DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC04504DA Supplier Thermo Scientific Chemicals Supplier No. CC04504DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4113-04-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
InChI Key VUAOIXANWIFYCU-UHFFFAOYSA-N
Synonym quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
PubChem CID 765653

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material 6-Quinolinecarbaldehyde