missing translation for 'onlineSavingsMsg'
Learn More

6-Piperidinopyridine-2-carboxylic acid, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 868755-50-4
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.25
MDL Number MFCD08435860
InChI Key ZBNWTBPSTOBOGV-UHFFFAOYSA-N
Synonym 6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
PubChem CID 18524421
IUPAC Name 6-piperidin-1-ylpyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(=CC=C1)N1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC38101CB
SDS View Documents
Thermo Scientific Chemicals
CC38101CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC38101DA
SDS View Documents
Thermo Scientific Chemicals
CC38101DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

868755-50-4
206.25
ZBNWTBPSTOBOGV-UHFFFAOYSA-N
18524421
OC(=O)C1=NC(=CC=C1)N1CCCCC1
C11H14N2O2
MFCD08435860
6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
6-piperidin-1-ylpyridine-2-carboxylic acid

Specifications

868755-50-4
100.0
Amber Glass Bottle
MFCD08435860
6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
OC(=O)C1=NC(=CC=C1)N1CCCCC1
206.25
206.24
6-Piperidinopyridine-2-carboxylic acid
96.5
97%
C11H14N2O2
250mg
ZBNWTBPSTOBOGV-UHFFFAOYSA-N
6-piperidin-1-ylpyridine-2-carboxylic acid
18524421
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents