missing translation for 'onlineSavingsMsg'
Learn More

6-Piperidinopyridine-2-carboxylic acid, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC38101DA

 View more versions of this product

Catalog No. CC38101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

6-Piperidinopyridine-2-carboxylic acid
96.5
97%
C11H14N2O2
1g
ZBNWTBPSTOBOGV-UHFFFAOYSA-N
6-(piperidin-1-yl)pyridine-2-carboxylic acid
18524421
97%
868755-50-4
100.0
Amber Glass Bottle
MFCD08435860
6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
OC(=O)C1=NC(=CC=C1)N1CCCCC1
206.25
206.24

Chemical Identifiers

868755-50-4
206.25
ZBNWTBPSTOBOGV-UHFFFAOYSA-N
18524421
OC(=O)C1=NC(=CC=C1)N1CCCCC1
C11H14N2O2
MFCD08435860
6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
6-(piperidin-1-yl)pyridine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions