missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenylpyridine-2-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America P23201G
Specifications
| 6-Phenylpyridine-2-carboxylic Acid | |
| 109°C | |
| C12H9NO2 | |
| 1 g | |
| CXOPUGLYEYDAMC-UHFFFAOYSA-N | |
| 6-phenylpyridine-2-carboxylic acid | |
| 2762884 | |
| ≥98.0% (GC,T) |
| 39774-28-2 | |
| White-Yellow | |
| MFCD04114206 | |
| 6-Phenylpicolinic Acid | |
| C1=CC=C(C=C1)C2=NC(=CC=C2)C(=O)O | |
| 199.209 | |
| 199.21 | |
| Crystalline Powder |
Chemical Identifiers
| 39774-28-2 | |
| 199.209 | |
| CXOPUGLYEYDAMC-UHFFFAOYSA-N | |
| 2762884 | |
| C1=CC=C(C=C1)C2=NC(=CC=C2)C(=O)O |
| C12H9NO2 | |
| MFCD04114206 | |
| 6-Phenylpicolinic Acid | |
| 6-phenylpyridine-2-carboxylic acid |
Safety and Handling
TSCA : No