missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenylpyridine-2-carboxylic Acid 98.0+%, TCI America™
$324.35 - $961.86
Chemical Identifiers
| CAS | 39774-28-2 |
|---|---|
| Molecular Formula | C12H9NO2 |
| Molecular Weight (g/mol) | 199.209 |
| MDL Number | MFCD04114206 |
| InChI Key | CXOPUGLYEYDAMC-UHFFFAOYSA-N |
| Synonym | 6-Phenylpicolinic Acid |
| PubChem CID | 2762884 |
| IUPAC Name | 6-phenylpyridine-2-carboxylic acid |
| SMILES | C1=CC=C(C=C1)C2=NC(=CC=C2)C(=O)O |
Chemical Identifiers
| 39774-28-2 | |
| 199.209 | |
| CXOPUGLYEYDAMC-UHFFFAOYSA-N | |
| 2762884 | |
| C1=CC=C(C=C1)C2=NC(=CC=C2)C(=O)O |
| C12H9NO2 | |
| MFCD04114206 | |
| 6-Phenylpicolinic Acid | |
| 6-phenylpyridine-2-carboxylic acid |
Specifications
| 39774-28-2 | |
| White-Yellow | |
| MFCD04114206 | |
| 6-Phenylpicolinic Acid | |
| C1=CC=C(C=C1)C2=NC(=CC=C2)C(=O)O | |
| 199.209 | |
| 199.21 | |
| Crystalline Powder |
| 109°C | |
| C12H9NO2 | |
| 1 g | |
| CXOPUGLYEYDAMC-UHFFFAOYSA-N | |
| 6-phenylpyridine-2-carboxylic acid | |
| 2762884 | |
| ≥98.0% (GC,T) | |
| 6-Phenylpyridine-2-carboxylic Acid |
Safety and Handling
TSCA : No