missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenylisocytosine 98.0+%, TCI America™
$195.73 - $587.17
Chemical Identifiers
| CAS | 56741-94-7 |
|---|---|
| Molecular Formula | C10H9N3O |
| Molecular Weight (g/mol) | 187.202 |
| MDL Number | MFCD00466412 |
| InChI Key | KXYGHDWFGFZJPJ-UHFFFAOYSA-N |
| Synonym | 2-Amino-6-phenyl-4(1H)-pyrimidinone |
| PubChem CID | 95891 |
| IUPAC Name | 2-amino-6-phenyl-1H-pyrimidin-4-one |
| SMILES | C1=CC=C(C=C1)C2=CC(=O)N=C(N2)N |
Chemical Identifiers
| 56741-94-7 | |
| 187.202 | |
| KXYGHDWFGFZJPJ-UHFFFAOYSA-N | |
| 95891 | |
| C1=CC=C(C=C1)C2=CC(=O)N=C(N2)N |
| C10H9N3O | |
| MFCD00466412 | |
| 2-Amino-6-phenyl-4(1H)-pyrimidinone | |
| 2-amino-6-phenyl-1H-pyrimidin-4-one |
Specifications
| 56741-94-7 | |
| White-Yellow | |
| MFCD00466412 | |
| 2-Amino-6-phenyl-4(1H)-pyrimidinone | |
| C1=CC=C(C=C1)C2=CC(=O)N=C(N2)N | |
| 187.202 | |
| 187.20 | |
| Crystalline Powder |
| 295°C | |
| C10H9N3O | |
| 1 g | |
| KXYGHDWFGFZJPJ-UHFFFAOYSA-N | |
| 2-amino-6-phenyl-1H-pyrimidin-4-one | |
| 95891 | |
| ≥98.0% (HPLC) | |
| 6-Phenylisocytosine |
Safety and Handling
TSCA : No