missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenyl-2-pyridone 98.0+%, TCI America™
$486.52 - $486.52
Chemical Identifiers
| CAS | 19006-82-7 |
|---|---|
| Molecular Formula | C11H9NO |
| Molecular Weight (g/mol) | 171.20 |
| MDL Number | MFCD00059760 |
| InChI Key | ZLUIDXRFWSPPCC-UHFFFAOYSA-N |
| PubChem CID | 87889 |
| IUPAC Name | 6-phenyl-1,2-dihydropyridin-2-one |
| SMILES | O=C1NC(=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 19006-82-7 | |
| 171.20 | |
| ZLUIDXRFWSPPCC-UHFFFAOYSA-N | |
| 6-phenyl-1,2-dihydropyridin-2-one |
| C11H9NO | |
| MFCD00059760 | |
| 87889 | |
| O=C1NC(=CC=C1)C1=CC=CC=C1 |
Specifications
| 19006-82-7 | |
| Yellow | |
| MFCD00059760 | |
| ZLUIDXRFWSPPCC-UHFFFAOYSA-N | |
| 6-phenyl-1,2-dihydropyridin-2-one | |
| 87889 | |
| ≥98.0% (GC,T) | |
| 6-Phenyl-2-pyridone |
| 197°C | |
| C11H9NO | |
| 1 g | |
| O=C1NC(=CC=C1)C1=CC=CC=C1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No