missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroveratraldehyde, 96%

Catalog No. p-7021892
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441506 5 g
AAA1441509 10 g
AAA1441514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1441506 Supplier Thermo Scientific Chemicals Supplier No. A1441506
Only null left

CAS: 20357-25-9 | C9H9NO5 | 211.17 g/mol

6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.

Solubility
Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.

Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 20357-25-9
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00007134
InChI Key YWSPWKXREVSQCA-UHFFFAOYSA-N
Synonym 6-nitroveratraldehyde, dmnb, 3,4-dimethoxy-6-nitrobenzaldehyde, 4,5-dimethoxy-2-nitro-benzaldehyde, dna-pk inhibitor, benzaldehyde, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzaldehyde, dna-dependent protein kinase inhibitor, 2-nitro-4,5-bis methyloxy benzaldehyde, pubchem8213
PubChem CID 88505
IUPAC Name 4,5-dimethoxy-2-nitrobenzaldehyde
SMILES COC1=CC(C=O)=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 131°C to 135°C
Quantity 5 g
Beilstein 395599
Sensitivity Air and light sensitive
Solubility Information Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.
Formula Weight 211.17
Percent Purity 96%
Chemical Name or Material 6-Nitroveratraldehyde
EINECSNumber 243-762-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only