missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindole 98.0+%, TCI America™

Catalog No. N04025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04025G 5 g
N04021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04025G Supplier TCI America Supplier No. N04025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4769-96-4
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00051497
InChI Key PSWCIARYGITEOY-UHFFFAOYSA-N
Synonym 6-nitroindole, 1h-indole, 6-nitro, 6-nitro-indole, 6-nitro-ih-indole, pubchem7242, #, acmc-1aq0p
PubChem CID 78502
IUPAC Name 6-nitro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2C=CNC2=C1

Specifications

Melting Point 143°C
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroindole