missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindole, 97%

Catalog No. AAA1292706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1292706 5 g
AAA1292703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1292706 Supplier Thermo Scientific Chemicals Supplier No. A1292706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4769-96-4 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4769-96-4
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00051497
InChI Key PSWCIARYGITEOY-UHFFFAOYSA-N
Synonym 6-nitroindole, 1h-indole, 6-nitro, 6-nitro-indole, 6-nitro-ih-indole, pubchem7242, #, acmc-1aq0p
PubChem CID 78502
IUPAC Name 6-nitro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2C=CNC2=C1

Specifications

Melting Point 141°C to 145°C
Quantity 5 g
Beilstein 135977
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 6-Nitroindole
Hazard Category H341
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 225-311-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only