missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrobenzothiazole, 98+%

CAS: 2942-06-5 | C7H4N2O2S | 180.18 g/mol

$80.85 - $538.42

Chemical Identifiers

CAS 2942-06-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.18
MDL Number MFCD00014569
InChI Key QLUFBCVWKTWKBF-UHFFFAOYSA-N
IUPAC Name 6-nitro-1,3-benzothiazole
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1063406
View Documents
Thermo Scientific Chemicals
A1063406
5 g
Each for $80.85
Only null left
 
AAA1063418
View Documents
Thermo Scientific Chemicals
A1063418
50 g
Each for $538.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2942-06-5
180.18
QLUFBCVWKTWKBF-UHFFFAOYSA-N
C7H4N2O2S
MFCD00014569
6-nitro-1,3-benzothiazole

Specifications

2942-06-5
C7H4N2O2S
5 g
5-nitrobenzothiazole, 5-nitrobenzo d thiazole, 5-nitro-benzothiazole, benzothiazole, 5-nitro, pubchem21844, 1,3-benzothiazole, 5-nitro, #, benzothiazole, 5-nitro-6ci,7ci,8ci,9ci
[O-][N+](=O)C1=CC=C2N=CSC2=C1
180.18
180.18
6-Nitrobenzothiazole
173°C to 177°C
MFCD00014569
7824
QLUFBCVWKTWKBF-UHFFFAOYSA-N
6-nitro-1,3-benzothiazole
350135
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-933-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only