missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitro-2,3-xylidine 98.0+%, TCI America™
$119.36 - $806.64
Chemical Identifiers
| CAS | 59146-96-2 |
|---|---|
| Molecular Formula | C8H10N2O2 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00010148 |
| InChI Key | YEFYPFWBLCARLC-UHFFFAOYSA-N |
| PubChem CID | 597166 |
| IUPAC Name | 2,3-dimethyl-6-nitroaniline |
| SMILES | CC1=C(C(=C(C=C1)[N+](=O)[O-])N)C |
Chemical Identifiers
| 59146-96-2 | |
| 166.18 | |
| YEFYPFWBLCARLC-UHFFFAOYSA-N | |
| 2,3-dimethyl-6-nitroaniline |
| C8H10N2O2 | |
| MFCD00010148 | |
| 597166 | |
| CC1=C(C(=C(C=C1)[N+](=O)[O-])N)C |
Specifications
| 118°C | |
| 25 g | |
| C8H10N2O2 | |
| 2811 | |
| YEFYPFWBLCARLC-UHFFFAOYSA-N | |
| 2,3-dimethyl-6-nitroaniline | |
| 597166 | |
| ≥98.0% (GC) |
| 59146-96-2 | |
| Crystal-Powder at 20°C | |
| MFCD00010148 | |
| 6-nitro-2,3-xylidine, 2,3-dimethyl-6-nitro-aniline, 2,3-dimethyl-6-nitrophenylamine, 4-nitro-3-aminoxylol, acmc-209mau, benzenamine,2,3-dimethyl-6-nitro, benzenamine, 2,3-dimethyl-6-nitro | |
| CC1=C(C(=C(C=C1)[N+](=O)[O-])N)C | |
| 166.18 | |
| 166.18 | |
| 6-Nitro-2,3-xylidine |
Safety and Handling
TSCA : No