missing translation for 'onlineSavingsMsg'
Learn More

BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo

6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo are miniature needles designed to provide highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.

$927.82 - $1256.82

Specifications

For Use With (Application) Gene and Vaccine Delivery
For Use With (Equipment) AgilePulse™ in Vivo Gene Delivery Electroporation System
Material Stainless Steel
Product Line BTX Electroporation
Sterility Sterile
View More Specs

Products 5
Catalog Number Mfr. No. Size (Metric) Gap Length (Metric) Tip Type Price Quantity  
Catalog Number Mfr. No. Size (Metric) Gap Length (Metric) Tip Type Price Quantity  
BTX470050
View Documents
BTX™
470050
4 mm 2 mm Fine Point
Each for $927.82
Only null left
 
BTX470060
View Documents
BTX™
470060
6 mm 2 mm Fine Point
Each for $927.82
Only null left
 
BTX470070
View Documents
BTX™
470070
6 mm 10 mm Fine Point
Each for $927.82
Only null left
 
BTX470086
View Documents
BTX™
470086
6 mm 16 mm Trocar Point
Each for $1,256.82
Only null left
 
BTX470080
View Documents
BTX™
470080
6 mm 25 mm Trocar Point
Each for $927.82
Only null left
 
Description

Description

The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.

The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.

  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from
Specifications

Specifications

Gene and Vaccine Delivery
Stainless Steel
Sterile
Square Wave
AgilePulse™ in Vivo Gene Delivery Electroporation System
BTX Electroporation
Electrode Connector Cable
Videos
SDS
Documents

Documents

Intended for Research use only. Not for use in diagnostic or clinical procedures.