missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
Specifications
| For Use With (Application) | Gene and Vaccine Delivery |
| For Use With (Equipment) | AgilePulse™ in Vivo Gene Delivery Electroporation System |
| Material | Stainless Steel |
| Product Line | BTX Electroporation |
| Product Type | Electroporation Needle Array |
| Description | 6-needle Array, 6 mm gap, 16 mm length |
| Diameter (English) | 0.03 in. |
| Diameter (Metric) | 0.7 mm |
| Electrode Shape | Straight/Needle array |
| Includes | 6-needle Array |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.