missing translation for 'onlineSavingsMsg'
Learn More

6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%

Catalog No. AAH27430MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27430MD 250 mg
AAH2743003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27430MD Supplier Thermo Scientific Chemicals Supplier No. H27430MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1032758-87-4 | C17H27BN2O4 | 334.223 g/mol

Used in the synthesis of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of organic compounds.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1032758-87-4
Molecular Formula C17H27BN2O4
Molecular Weight (g/mol) 334.223
MDL Number MFCD09037484
InChI Key QGBJKHALDLKAGX-UHFFFAOYSA-N
Synonym tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-boc-methylamino pyridine-3-boronic acid pinacol ester, 6-n-boc-methylamino pyridine-3-boronic acid pinacol ester, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-tert-butoxycarbonyl methyl amino pyridin-3-yl boronic acid pinacol ester, tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-yl carbamate, tert-butylmethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
PubChem CID 44541676
IUPAC Name tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N(C)C(=O)OC(C)(C)C

Specifications

Melting Point 112°C to 119°C
Quantity 250 mg
Formula Weight 334.22
Percent Purity 95%
Chemical Name or Material 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only