accessibility menu, dialog, popup

UserName

CAS RN 1032758-87-4

IUPAC Name: tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 334.22
Molecular Formula: C17H27BN2O4
InChi Key: QGBJKHALDLKAGX-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results