missing translation for 'onlineSavingsMsg'
Learn More

6-Methylsalicylic acid, 98%

Catalog No. p-7077403
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453060050 5 g
AC453060250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453060050 Supplier Thermo Scientific Chemicals Supplier No. 453060050
Only null left

CAS: 567-61-3 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 567-61-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD01194284
InChI Key HCJMNOSIAGSZBM-UHFFFAOYSA-N
Synonym 6-methylsalicylic acid, 6-hydroxy-o-toluic acid, 2,6-cresotic acid, 6-msa, benzoic acid, 2-hydroxy-6-methyl, 6-ms, methylsalicylic acid, 2-hydroxy-6-methylbenzoicacid, unii-l5352fe23y, 2-carboxy-3-methylphenol
PubChem CID 11279
ChEBI CHEBI:17637
IUPAC Name 2-hydroxy-6-methylbenzoic acid
SMILES CC1=CC=CC(O)=C1C(O)=O

Specifications

Melting Point 170.0°C to 175.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 6-Methylsalicylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only