missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-3-carboxaldehyde, 96%

Catalog No. AAH66926MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66926MD 250 mg
AAH6692603 1 g
AAH6692606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66926MD Supplier Thermo Scientific Chemicals Supplier No. H66926MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53014-84-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53014-84-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD08272279
InChI Key IMWMEIWYPWVABQ-UHFFFAOYSA-N
Synonym 6-methylnicotinaldehyde, 2-methyl-5-formylpyridine, 6-methyl-pyridine-3-carbaldehyde, 3-formyl-6-methyl-pyridine, 5-formyl-2-methylpyridine, 5-formyl-2-picoline, 6-methylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methyl, 3-formyl-6-methylpyridine, 6-methylpyridyl-3-carboxaldehyde
PubChem CID 10192566
IUPAC Name 6-methylpyridine-3-carbaldehyde
SMILES CC1=NC=C(C=O)C=C1

Specifications

Density 1.084 g/mL
Boiling Point 95°C (20 mmHg)
Flash Point 87°C (189°F)
Refractive Index 1.546
Quantity 250 mg
Beilstein 107496
Sensitivity Moisture sensitive
Formula Weight 121.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 6-Methylpyridine-3-carboxaldehyde
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only