accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 53014-84-9

IUPAC Name: 6-methylpyridine-3-carbaldehyde
Synonyms: 6-methylpyridine-3-carbaldehyde
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: IMWMEIWYPWVABQ-UHFFFAOYSA-N
SMILES: CC1=NC=C(C=O)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results