missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 6-α-Methylprednisolone

Supplier:  Thermo Scientific™ 338800050

 View more versions of this product

Catalog No. AC338800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-α-Methylprednisolone
C22H30O5
5g
methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol
VHRSUDSXCMQTMA-PJHHCJLFSA-N
(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
6741
374.48
83-43-2
MFCD00010591
15,6184
Solubility in water: insoluble. Other solubilities: slightly soluble in acetone
C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12
374.48
CHEBI:6888

Chemical Identifiers

83-43-2
374.48
VHRSUDSXCMQTMA-PJHHCJLFSA-N
6741
(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
C22H30O5
MFCD00010591
methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol
CHEBI:6888
C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 201-476-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only