missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinamide 98.0+%, TCI America™

Catalog No. p-7133243
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24775G 5 g
M247725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24775G Supplier TCI America Supplier No. M24775G

Chemical Identifiers

CAS 6960-22-1
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00006342
InChI Key IJXDURUAYOKSIS-UHFFFAOYSA-N
Synonym 6-methylnicotinamide, 6-methyl nicotinamide, 3-pyridinecarboxamide, 6-methyl, 6-methyl-3-pyridinecarboxamide, 8ej, nicotinamide, 6-methyl, 3-carbamoyl-6-methylpyridine, ksc630k7f, 2-methylpyridine-5-carboxamide, 3-pyridinecarboxamide,6-methyl
PubChem CID 96351
IUPAC Name 6-methylpyridine-3-carboxamide
SMILES CC1=NC=C(C=C1)C(=O)N

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylnicotinamide