accessibility menu, dialog, popup

UserName

CH3NNH2O

CAS RN 6960-22-1

IUPAC Name: 6-methylpyridine-3-carboxamide
Synonyms: 6-methylpyridine-3-carboxamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: IJXDURUAYOKSIS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=N1)C(N)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)