missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinamide, 98%

Catalog No. AAA1603903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1603903 1 g
AAA1603906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1603903 Supplier Thermo Scientific Chemicals Supplier No. A1603903

CAS: 6960-22-1 | C7H8N2O | 136.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6960-22-1
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00006342
InChI Key IJXDURUAYOKSIS-UHFFFAOYSA-N
PubChem CID 96351
IUPAC Name 6-methylpyridine-3-carboxamide
SMILES CC1=NC=C(C=C1)C(=O)N

Specifications

Melting Point 195°C to 199°C
Quantity 1 g
Beilstein 112050
Synonym 6-methylnicotinamide, 6-methyl nicotinamide, 3-pyridinecarboxamide, 6-methyl, 6-methyl-3-pyridinecarboxamide, 8ej, nicotinamide, 6-methyl, 3-carbamoyl-6-methylpyridine, ksc630k7f, 2-methylpyridine-5-carboxamide, 3-pyridinecarboxamide,6-methyl
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 6-Methylnicotinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only