missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylindole 98.0+%, TCI America™
Supplier: TCI America M14305G
Specifications
| 6-Methylindole | |
| C9H9N | |
| 5 g | |
| ONYNOPPOVKYGRS-UHFFFAOYSA-N | |
| 6-methyl-1H-indole | |
| 137928 | |
| ≥98.0% (GC) |
| 3420-02-8 | |
| MFCD00005682 | |
| 6-methylindole, 1h-indole, 6-methyl, 6-methyl indole, indole, 6-methyl, pubchem7240, 6-methyl-indole, acmc-1cokr, #, ksc491i2r | |
| CC1=CC2=C(C=C1)C=CN2 | |
| 131.18 | |
| 131.18 | |
| Crystalline Lumps |
Chemical Identifiers
| 3420-02-8 | |
| 131.18 | |
| ONYNOPPOVKYGRS-UHFFFAOYSA-N | |
| 137928 | |
| CC1=CC2=C(C=C1)C=CN2 |
| C9H9N | |
| MFCD00005682 | |
| 6-methylindole, 1h-indole, 6-methyl, 6-methyl indole, indole, 6-methyl, pubchem7240, 6-methyl-indole, acmc-1cokr, #, ksc491i2r | |
| 6-methyl-1H-indole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator