missing translation for 'onlineSavingsMsg'
Learn More

6-Methylindole 98.0+%, TCI America™

Catalog No. p-7132727
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14301G 1 g
M14305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14301G Supplier TCI America Supplier No. M14301G
Only null left

Chemical Identifiers

CAS 3420-02-8
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005682
InChI Key ONYNOPPOVKYGRS-UHFFFAOYSA-N
Synonym 6-methylindole, 1h-indole, 6-methyl, 6-methyl indole, indole, 6-methyl, pubchem7240, 6-methyl-indole, acmc-1cokr, #, ksc491i2r
PubChem CID 137928
IUPAC Name 6-methyl-1H-indole
SMILES CC1=CC2=C(C=C1)C=CN2

Specifications

Quantity 1 g
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methylindole
TSCA No
Recommended Storage Refrigerator