missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-nitrouracil 95.0+%, TCI America™

Catalog No. M25065G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25065G 5 g
M25061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25065G Supplier TCI America Supplier No. M25065G
Only null left

Chemical Identifiers

CAS 16632-21-6
Molecular Formula C5H5N3O4
Molecular Weight (g/mol) 171.11
MDL Number MFCD00051201
InChI Key LIVYMRJSNFHYEN-UHFFFAOYSA-N
Synonym 5-nitro-6-methyluracil, 6-methyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4 1h,3h-dione, 2,4-dihydroxy-6-methyl-5-nitropyrimidine, 6-metyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4-diol, uracil, 6-methyl-5-nitro, 2,4 1h,3h-pyrimidinedione, 6-methyl-5-nitro, 4-methyl-5-nitrouracil, 2,4-dihydroxy-6-methyl-5-*nitropyrimidine
PubChem CID 85523
IUPAC Name 6-methyl-5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=C(C(=O)NC(=O)N1)[N+]([O-])=O

Specifications

Melting Point 276°C
Color Yellow
Quantity 5 g
Formula Weight 171.11
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-5-nitrouracil