missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-4-chromanone 98.0+%, TCI America™
$100.11 - $290.70
Chemical Identifiers
| CAS | 39513-75-2 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.19 |
| MDL Number | MFCD00024071 |
| InChI Key | RJHXEPLSJAVTFW-UHFFFAOYSA-N |
| PubChem CID | 584354 |
| IUPAC Name | 6-methyl-3,4-dihydro-2H-1-benzopyran-4-one |
| SMILES | CC1=CC2=C(OCCC2=O)C=C1 |
Chemical Identifiers
| 39513-75-2 | |
| 162.19 | |
| RJHXEPLSJAVTFW-UHFFFAOYSA-N | |
| 6-methyl-3,4-dihydro-2H-1-benzopyran-4-one |
| C10H10O2 | |
| MFCD00024071 | |
| 584354 | |
| CC1=CC2=C(OCCC2=O)C=C1 |
Specifications
| 39513-75-2 | |
| White-Yellow | |
| C10H10O2 | |
| 1 g | |
| CC1=CC2=C(OCCC2=O)C=C1 | |
| 162.19 | |
| 162.19 | |
| Crystalline Lumps |
| 35°C | |
| 162°C | |
| MFCD00024071 | |
| RJHXEPLSJAVTFW-UHFFFAOYSA-N | |
| 6-methyl-3,4-dihydro-2H-1-benzopyran-4-one | |
| 584354 | |
| ≥98.0% (GC) | |
| 6-Methyl-4-chromanone |
Safety and Handling
TSCA : No