accessibility menu, dialog, popup

UserName

H3COO

CAS RN 39513-75-2

IUPAC Name: 6-methyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 6-methyl-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: RJHXEPLSJAVTFW-UHFFFAOYSA-N
SMILES: CC1=CC2=C(OCCC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results