missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-4-chromanone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 35°C |
| Color | White-Yellow |
| Boiling Point | 162°C |
| Quantity | 1 g |
| Formula Weight | 162.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 6-Methyl-4-chromanone |
Safety and Handling
| TSCA | No |