missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3-pyridinemethanol 97.0+%, TCI America™

Catalog No. p-7133368
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26831G 1 g
M26835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26831G Supplier TCI America Supplier No. M26831G
Only null left

Chemical Identifiers

CAS 34107-46-5
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD08692003
InChI Key DJCJOWDAAZEMCI-UHFFFAOYSA-N
Synonym 5-Hydroxymethyl-2-methylpyridine, 5-Hydroxymethyl-2-picoline
PubChem CID 11228788
IUPAC Name (6-methylpyridin-3-yl)methanol
SMILES CC1=NC=C(C=C1)CO

Specifications

Melting Point 46°C
Boiling Point 100°C
Quantity 1 g
Formula Weight 123.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methyl-3-pyridinemethanol