accessibility menu, dialog, popup

UserName

CH3NOH

CAS RN 34107-46-5

IUPAC Name: (6-methylpyridin-3-yl)methanol
Synonyms: (6-methylpyridin-3-yl)methanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: DJCJOWDAAZEMCI-UHFFFAOYSA-N
SMILES: CC1=NC=C(CO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results