missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one 98.0+%, TCI America™

Catalog No. m11165g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11165G Supplier TCI America Supplier No. M11165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16075-68-6
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.176
MDL Number MFCD00191466
InChI Key XVVOALIRSDLWJA-UHFFFAOYSA-N
PubChem CID 21766687
IUPAC Name 6-methylpyrido[1,2-a]pyrimidin-2-one
SMILES CC1=CC=CC2=NC(=O)C=CN12

Specifications

Quantity 5 g
Formula Weight 160.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one