missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-thiouracil 98.0+%, TCI America™

Catalog No. p-7132175
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044325G 25 g
M0443500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M044325G Supplier TCI America Supplier No. M044325G

Chemical Identifiers

CAS 56-04-2
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00006040
InChI Key HWGBHCRJGXAGEU-UHFFFAOYSA-N
Synonym methylthiouracil, 6-methyl-2-thiouracil, methiocil, 4-hydroxy-2-mercapto-6-methylpyrimidine, thimecil, thiomecil, thiothymin, alkiron, metacil, 2-mercapto-6-methylpyrimidin-4-ol
PubChem CID 667493
ChEBI CHEBI:82346
IUPAC Name 6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES CC1=CC(=O)NC(=S)N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 142.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-2-thiouracil
EINECSNumber (5)-0935
RTECSNumber YR0875000
TSCA Yes