Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Methyl-2-thiouracil 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 142.18 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Methyl-2-thiouracil |
Safety and Handling
| EINECSNumber | (5)-0935 |
| RTECSNumber | YR0875000 |
| TSCA | Yes |