missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-thiouracil, 98%

Catalog No. AAA1798218
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1798218 50 g
AAA1798230 250 g
AAA17982A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1798218 Supplier Thermo Scientific Chemicals Supplier No. A1798218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56-04-2 | C5H6N2OS | 142.18 g/mol

6-Methyl-2-thiouracil is mainly used in surgical preparation of mild hyperthyroidism, thyroid storm, hyperthyroidism and postoperative treatment. It is used in the treatment of hyperthyroidism. It is used as a drug, which is applied to decrease the formation of stored thyroid hormone and thyroglobulin in the thyroid gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-2-thiouracil is mainly used in surgical preparation of mild hyperthyroidism, thyroid storm, hyperthyroidism and postoperative treatment. It is used in the treatment of hyperthyroidism. It is used as a drug, which is applied to decrease the formation of stored thyroid hormone and thyroglobulin in the thyroid gland.

Solubility
Soluble in ethanol. Slightly soluble in methanol, acetone, diethyl ether, water, ammonia and alkali hydroxides. Insoluble in benzene and chloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Keep the container tightly closed.

Chemical Identifiers

CAS 56-04-2
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00006040
InChI Key HWGBHCRJGXAGEU-UHFFFAOYSA-N
Synonym methylthiouracil, 6-methyl-2-thiouracil, methiocil, 4-hydroxy-2-mercapto-6-methylpyrimidine, thimecil, thiomecil, thiothymin, alkiron, metacil, 2-mercapto-6-methylpyrimidin-4-ol
PubChem CID 667493
ChEBI CHEBI:82346
IUPAC Name 6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CC1=CC(=O)NC(=S)N1

Specifications

Melting Point ∼329°C (decomposition)
Quantity 50 g
Beilstein 115648
Merck Index 14,6128
Solubility Information Soluble in ethanol. Slightly soluble in methanol,acetone,diethyl ether,water,ammonia and alkali hydroxides. Insoluble in benzene and chloroform.
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-thiouracil
Hazard Category H302-H351
Hazard Statement GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-252-3
RTECSNumber YR0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only