missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-1,2,3,4-tetrahydroquinoline 97.0+%, TCI America™
$132.35 - $132.35
Chemical Identifiers
| CAS | 91-61-2 |
|---|---|
| Molecular Formula | C10H13N |
| Molecular Weight (g/mol) | 147.22 |
| MDL Number | MFCD00023887 |
| InChI Key | XOKMRXSMOHCNIX-UHFFFAOYSA-N |
| Synonym | civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa |
| PubChem CID | 66678 |
| IUPAC Name | 6-methyl-1,2,3,4-tetrahydroquinoline |
| SMILES | CC1=CC=C2NCCCC2=C1 |
Chemical Identifiers
| 91-61-2 | |
| 147.22 | |
| XOKMRXSMOHCNIX-UHFFFAOYSA-N | |
| 66678 | |
| CC1=CC=C2NCCCC2=C1 |
| C10H13N | |
| MFCD00023887 | |
| civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa | |
| 6-methyl-1,2,3,4-tetrahydroquinoline |
Specifications
| 91-61-2 | |
| Yellow | |
| MFCD00023887 | |
| civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa | |
| CC1=CC=C2NCCCC2=C1 | |
| 147.22 | |
| 147.22 | |
| Crystalline Lumps |
| 35°C | |
| C10H13N | |
| 5 g | |
| XOKMRXSMOHCNIX-UHFFFAOYSA-N | |
| 6-methyl-1,2,3,4-tetrahydroquinoline | |
| 66678 | |
| ≥97.0% (GC) | |
| 6-Methyl-1,2,3,4-tetrahydroquinoline |
Safety and Handling
EINECSNumber : (5)-0803
TSCA : Yes