missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypurine 98.0+%, TCI America™

Catalog No. p-7132915
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19251G 1 g
M19255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19251G Supplier TCI America Supplier No. M19251G
Only null left

Chemical Identifiers

CAS 1074-89-1
Molecular Formula C6H6N4O
Molecular Weight (g/mol) 150.14
MDL Number MFCD00127913
InChI Key GOILPRCCOREWQE-UHFFFAOYSA-N
Synonym 6-methoxypurine, 6-methoxy-9h-purine, 1h-purine, 6-methoxy, o-methylhypoxanthine, purine, 6-methoxy, 1h-purine, 6-methoxy-9ci, 6-methoxy-1h-purine, 6-methoxy purine, 6-methoxypu-rine, 6-methoxypurine 99+%
PubChem CID 70632
IUPAC Name 6-methoxy-7H-purine
SMILES COC1=C2NC=NC2=NC=N1

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 1 g
Formula Weight 150.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxypurine