accessibility menu, dialog, popup

UserName

Découvrez nos nouvelles offres vedettes sur l’équipement, les instruments et plus encore

Économisez maintenant

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1074-89-1

H3CONNNHN

CAS RN 1074-89-1

IUPAC Name: 6-methoxy-7H-purine
Synonyms: 6-methoxy-7H-purine
Molecular Weight (g/mol): 150.14
Molecular Formula: C6H6N4O
InChi Key: GOILPRCCOREWQE-UHFFFAOYSA-N
SMILES: COC1=C2NC=NC2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
1x1 image pixel for data collection