missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxy-4-methylquinoline Hydrate 98.0+%, TCI America™
$236.73 - $948.81
Chemical Identifiers
| CAS | 41037-26-7 |
|---|---|
| Molecular Formula | C11H11NO |
| Molecular Weight (g/mol) | 173.215 |
| MDL Number | MFCD00191547 |
| InChI Key | MBVGYFIYXWVPQY-UHFFFAOYSA-N |
| PubChem CID | 258761 |
| IUPAC Name | 6-methoxy-4-methylquinoline |
| SMILES | CC1=C2C=C(C=CC2=NC=C1)OC |
Chemical Identifiers
| 41037-26-7 | |
| 173.215 | |
| MBVGYFIYXWVPQY-UHFFFAOYSA-N | |
| 6-methoxy-4-methylquinoline |
| C11H11NO | |
| MFCD00191547 | |
| 258761 | |
| CC1=C2C=C(C=CC2=NC=C1)OC |
Specifications
| 41037-26-7 | |
| White-Yellow | |
| C11H11NO | |
| 1 g | |
| CC1=C2C=C(C=CC2=NC=C1)OC | |
| 173.215 | |
| 173.22 | |
| Crystalline Powder |
| 53°C | |
| 158°C | |
| MFCD00191547 | |
| MBVGYFIYXWVPQY-UHFFFAOYSA-N | |
| 6-methoxy-4-methylquinoline | |
| 258761 | |
| ≥98.0% (GC,T) | |
| 6-Methoxy-4-methylquinoline Hydrate |
Safety and Handling
TSCA : No